CH6371379104 Chem Impex International, Inc.

Na-Boc-Ne-Fmoc-D-lysine

  • MDL number MFCD00076966
  • Cas Number : 115186-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-140
Synonym Boc-D-Lys(Fmoc)-OH
Purity 99
Storage temp. Store at = -4
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-fmoc-d-lysine/3596?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-fmoc-d-lysine/3596?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart