CH6371379107 Chem Impex International, Inc.

Boc-D-Met-Gly-OH

  • MDL number MFCD00190812
  • Cas Number : 69612-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-131° C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-met-gly-oh/3599?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-met-gly-oh/3599?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart