CH6371379108 Chem Impex International, Inc.

Boc-D-methionine N-hydroxysuccinimide ester

  • MDL number MFCD00236867
  • Cas Number : 26060-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Met-OSu
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-methionine-i-n-i-hydroxysuccinimide-ester/3600?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-methionine-i-n-i-hydroxysuccinimide-ester/3600?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart