CH6371379109 Chem Impex International, Inc.

Na,d-Bis-Boc-D-ornithine

  • MDL number MFCD00151881
  • Cas Number : 137524-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-123
Synonym Boc-D-Orn(Boc)-OH
Purity 0.99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-delta-sup-bis-boc-d-ornithine/3601?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-delta-sup-bis-boc-d-ornithine/3601?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart