CH6371379110 Chem Impex International, Inc.

Na-Boc-Nd-Fmoc-D-ornithine

  • MDL number MFCD00151883
  • Cas Number : 163336-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-152
Synonym Boc-D-Orn(Fmoc)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-delta-sup-fmoc-d-ornithine/3602?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-delta-sup-fmoc-d-ornithine/3602?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart