CH6371379114 Chem Impex International, Inc.

Boc-D-Phe-Pro-OH

  • MDL number MFCD00237546
  • Cas Number : 38675-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-phe-pro-oh/3606?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-phe-pro-oh/3606?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart