CH6371379115 Chem Impex International, Inc.

Boc-O-tert-butyl-D-serine dicyclohexylammonium salt

  • MDL number MFCD00236872
  • Cas Number : 248921-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Ser(tBu)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-i-tert-i-butyl-d-serine-dicyclohexylammonium-salt/3607?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-i-tert-i-butyl-d-serine-dicyclohexylammonium-salt/3607?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart