CH6371379121 Chem Impex International, Inc.

Boc-O-2,6-dichlorobenzyl-DL-tyrosine

  • MDL number MFCD00236878
  • Cas Number : 201334-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-DL-Tyr(2,6-di-Cl-Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-dl-tyrosine/3613?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-dl-tyrosine/3613?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart