CH6371379123 Chem Impex International, Inc.

Boc-L-glutamic acid gamma-methyl ester dicyclohexylammonium salt

  • MDL number MFCD00076932
  • Cas Number : 14406-17-8
  • Molecular Weight : 442.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-159° C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-methyl-ester-dicyclohexylammonium-salt/3616?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-methyl-ester-dicyclohexylammonium-salt/3616?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart