CH6371379124 Chem Impex International, Inc.

Boc-L-glutamic acid g-methyl ester a-4-nitrophenyl ester

  • MDL number MFCD00190793
  • Cas Number : 16947-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-91
Synonym Boc-L-Glu(OMe)-ONp
Purity 99
Storage temp. Store at 0-8
Physical Form White powder with a hint of yellow
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-methyl-ester-a-4-nitrophenyl-ester/3617?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-methyl-ester-a-4-nitrophenyl-ester/3617?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart