CH6371379126 Chem Impex International, Inc.

Boc-L-glutamic acid g-N-hydroxysuccinimide ester a-tert-butyl ester

  • MDL number MFCD00236848
  • Cas Number : 81659-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-142
Synonym Boc-L-Glu(OSu)-OtBu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-gamma-i-n-i-hydroxysuccinimide-ester-alpha-i-tert-i-butyl-ester/3619?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-gamma-i-n-i-hydroxysuccinimide-ester-alpha-i-tert-i-butyl-ester/3619?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart