CH6371379127 Chem Impex International, Inc.

Boc-N-p-benzyloxymethyl-L-histidine methyl ester hydrochloride

  • MDL number MFCD00076941
  • Cas Number : 83468-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-143 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-p-benzyloxymethyl-l-histidine-methyl-ester-hydrochloride/3622?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-p-benzyloxymethyl-l-histidine-methyl-ester-hydrochloride/3622?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart