CH6371379128 Chem Impex International, Inc.

Boc-His-Met-OH

  • MDL number MFCD00237557
  • Cas Number : 248921-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-195° C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-his-met-oh/3625?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-his-met-oh/3625?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart