CH6371379130 Chem Impex International, Inc.

Boc-L-glutaminol

  • MDL number MFCD00236849
  • Cas Number : 133565-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-121° C (Crystals from ethanol-ether)
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutaminol/3628?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutaminol/3628?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart