CH6371379134 Chem Impex International, Inc.

Boc-Lys(Boc)-Gly-OH

  • MDL number MFCD00237560
  • Cas Number : 120893-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-lys-boc-gly-oh/3632?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-lys-boc-gly-oh/3632?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart