CH6371379135 Chem Impex International, Inc.

Na-Boc-Ne-4-toluenesulfonyl-L-lysine 4-nitrophenyl ester

  • MDL number MFCD00236859
  • Cas Number : 200943-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-115
Synonym Boc-L-Lys(Tos)-ONp
Purity 99
Storage temp. Store at 0-8
Physical Form Yellowish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-4-toluenesulfonyl-l-lysine-4-nitrophenyl-ester/3633?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-4-toluenesulfonyl-l-lysine-4-nitrophenyl-ester/3633?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart