CH6371379137 Chem Impex International, Inc.

Boc-N-ethyl-L-leucine dicyclohexylammonium salt

  • MDL number MFCD00237566
  • Cas Number : 200936-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-ethyl-l-leucine-dicyclohexylammonium-salt/3635?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-ethyl-l-leucine-dicyclohexylammonium-salt/3635?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart