CH6371379138 Chem Impex International, Inc.

Boc-N-methyl-2-aminobenzoic acid

  • MDL number MFCD00237567
  • Cas Number : 141871-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-2-aminobenzoic-acid/3636?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-2-aminobenzoic-acid/3636?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart