CH6371379140 Chem Impex International, Inc.

Boc-Na-methyl-Nw-(4-methoxy-2,3,6-trimethylbenzenesulfonyl)-L-arginine

  • MDL number MFCD00236863
  • Cas Number : 125602-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-N-Me-L-Arg(Mtr)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-sup-alpha-sup-methyl-i-n-i-sup-omega-sup-4-methoxy-2-3-6-trimethylbenzenesulfonyl-l-arginine/3638?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-sup-alpha-sup-methyl-i-n-i-sup-omega-sup-4-methoxy-2-3-6-trimethylbenzenesulfonyl-l-arginine/3638?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart