CH6371379141 Chem Impex International, Inc.

Na-Boc-Na-methyl-Nw-4-toluenesulfonyl-L-arginine

  • MDL number MFCD00134858
  • Cas Number : 108695-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-136� C
Synonym Boc-Nalpha-methyl-Nomega-4-toluenesulfonyl-L-arginine
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-a-sup-methyl-i-n-i-sup-sup-4-toluenesulfonyl-l-arginine/3639?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-a-sup-methyl-i-n-i-sup-sup-4-toluenesulfonyl-l-arginine/3639?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart