CH6371379143 Chem Impex International, Inc.

Boc-N-methyl-D-isoleucine

  • MDL number MFCD00236865
  • Cas Number : 267223-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-56° C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-d-isoleucine/3642?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-d-isoleucine/3642?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart