CH6371379145 Chem Impex International, Inc.

Boc-N-methyl-DL-valine

  • MDL number MFCD00153317
  • Cas Number : 13850-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-88� C
Synonym Boc-N-Me-DL-Val-OH
Purity 0.98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-dl-valine/3644?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-dl-valine/3644?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart