CH6371379149 Chem Impex International, Inc.

Boc-N-methyl-4-nitro-L-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD00038840
  • Cas Number : 70663-56-8
  • Molecular Weight : 505.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-4-nitro-l-phenylalanine-dicyclohexylammonium-salt/3648?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-4-nitro-l-phenylalanine-dicyclohexylammonium-salt/3648?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart