CH6371379152 Chem Impex International, Inc.

Boc-N-methyl-O-acetyl-L-tyrosine dicyclohexylammonium salt

  • MDL number MFCD00236868
  • Cas Number : 201294-70-4
  • Molecular Weight : 518.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-157° C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-acetyl-l-tyrosine-dicyclohexylammonium-salt/3651?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-i-o-i-acetyl-l-tyrosine-dicyclohexylammonium-salt/3651?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart