CH6371379157 Chem Impex International, Inc.

Boc-4-iodo-DL-phenylalanine

  • MDL number MFCD00176869
  • Cas Number : 103882-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-169� C
Synonym Boc-DL-Phe(4-I)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-iodo-dl-phenylalanine/3656?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-iodo-dl-phenylalanine/3656?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart