CH6371379160 Chem Impex International, Inc.

Boc-Phe-Phe-OH

  • MDL number MFCD00190824
  • Cas Number : 13122-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133 - 136
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-phe-phe-oh/3659?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-phe-phe-oh/3659?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart