CH6371379165 Chem Impex International, Inc.

Boc-L-pyroglutamic acid dicyclohexylammonium salt

  • MDL number MFCD00076982
  • Cas Number : 4677-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-201
Synonym Boc-L-Pyr-OH DCHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-pyroglutamic-acid-dicyclohexylammonium-salt/3665?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-pyroglutamic-acid-dicyclohexylammonium-salt/3665?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart