CH6371379171 Chem Impex International, Inc.

Boc-b-chloro-L-alanine

  • MDL number MFCD00672318
  • Cas Number : 71404-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-beta-chloro-L-Ala-OH
Purity 95
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-beta-chloro-l-alanine/3671?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-beta-chloro-l-alanine/3671?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart