CH6371379172 Chem Impex International, Inc.

Boc-beta-cyano-L-alanine

  • MDL number MFCD00237539
  • Cas Number : 45159-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-ß-cyano-l-alanine/3672?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-ß-cyano-l-alanine/3672?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart