CH6371379173 Chem Impex International, Inc.

Boc-b-cyano-D-alanine

  • MDL number MFCD00236830
  • Cas Number : 184685-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-b-cyano-D-Ala-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-beta-cyano-d-alanine/3673?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-beta-cyano-d-alanine/3673?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart