CH6371379178 Chem Impex International, Inc.

Boc-O-4-chlorobenzyl-L-threonine dicyclohexylammonium salt

  • MDL number MFCD00076989

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-156 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-4-chlorobenzyl-l-threonine-dicyclohexylammonium-salt/3678?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-4-chlorobenzyl-l-threonine-dicyclohexylammonium-salt/3678?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart