CH6371379179 Chem Impex International, Inc.

Boc-O-acetyl-L-tyrosine

  • MDL number MFCD00057973
  • Cas Number : 80971-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 85 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-acetyl-l-tyrosine/3679?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-acetyl-l-tyrosine/3679?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart