CH6371379183 Chem Impex International, Inc.

Bz-Arg-4-Abz-OH�

  • MDL number MFCD00082418

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Nalpha-Benzoyl-L-arginine 4-carboxyanilide hydrochloride
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/bz-arg-4-abz-oh·hcl/3683?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/bz-arg-4-abz-oh·hcl/3683?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart