CH6371379186 Chem Impex International, Inc.

Na-Benzoyl-DL-arginine hydrochloride

  • MDL number MFCD06795766
  • Cas Number : 125652-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bz-DL-Arg-OH HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-benzoyl-dl-arginine-hydrochloride/3691?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-benzoyl-dl-arginine-hydrochloride/3691?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart