CH6371379189 Chem Impex International, Inc.

Benzoyl-DL-phenylalanine beta-naphthylamide

  • MDL number MFCD00021596
  • Cas Number : 100900-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-141 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White product
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-dl-phenylalanine-beta-naphthylamide/3696?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-dl-phenylalanine-beta-naphthylamide/3696?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart