CH6371379191 Chem Impex International, Inc.

Benzoyl-L-Valine

  • MDL number MFCD00066064
  • Cas Number : 5699-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-137� C
Synonym Bz-L-Val-OH
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-l-valine/3699?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-l-valine/3699?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart