CH6371379194 Chem Impex International, Inc.

Na-Benzyl-L-histidine methyl ester dihydrochloride

  • MDL number MFCD00167636
  • Cas Number : 102029-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bzl-L-His-OMe 2HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzyl-l-histidine-methyl-ester-dihydrochloride/3703?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzyl-l-histidine-methyl-ester-dihydrochloride/3703?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart