CH6371379195 Chem Impex International, Inc.

Benzyl-L-isoleucine

  • MDL number MFCD00038331
  • Cas Number : 1859-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219-223� c
Synonym Bzl-L-Ile-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-l-isoleucine/3704?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-l-isoleucine/3704?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart