CH6371379196 Chem Impex International, Inc.

N-Benzyl-N-methyl-L-serine

  • MDL number MFCD00237598
  • Cas Number : 201208-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bzl-N-Me-L-Ser-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-i-n-i-methyl-l-serine/3705?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-i-n-i-methyl-l-serine/3705?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart