CH6371379202 Chem Impex International, Inc.

DL-a-Aminosuberic acid

  • MDL number MFCD00063122
  • Cas Number : 3054-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Asu-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-a-aminosuberic-acid/3711?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-a-aminosuberic-acid/3711?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart