CH6371379203 Chem Impex International, Inc.

DL-2-Acetylamino-6-N-Boc-amino-4-hexynoic acid�A

  • MDL number MFCD00236749
  • Cas Number : 90102-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-2-acetylamino-6-n-boc-amino-4-hexynoic-acid·dcha/3713?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-2-acetylamino-6-n-boc-amino-4-hexynoic-acid·dcha/3713?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart