CH6371379204 Chem Impex International, Inc.

DL-alpha-Aminopimelic acid

  • MDL number MFCD00053314
  • Cas Number : 627-76-9
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-216 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-a-aminopimelic-acid/3714?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-a-aminopimelic-acid/3714?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart