CH6371379208 Chem Impex International, Inc.

Na-Fmoc-L-asparagine 4-nitrophenyl ester

  • MDL number MFCD00063329
  • Cas Number : 71989-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-188� C
Synonym Fmoc-L-Asn-ONp
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-l-asparagine-4-nitrophenyl-ester/3727?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-l-asparagine-4-nitrophenyl-ester/3727?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart