CH6371379211 Chem Impex International, Inc.

Fmoc-L-aspartic acid a-benzyl ester

  • MDL number MFCD00198201
  • Cas Number : 86060-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Asp-OBzl
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-aspartic-acid-a-benzyl-ester/3730?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-aspartic-acid-a-benzyl-ester/3730?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart