CH6371379213 Chem Impex International, Inc.

Fmoc-S-2-hydroxyethyl-L-cysteine

  • MDL number MFCD00672330
  • Cas Number : 200354-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Cys(2-hydroxyethyl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-2-hydroxyethyl-l-cysteine/3732?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-2-hydroxyethyl-l-cysteine/3732?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart