CH6371379215 Chem Impex International, Inc.

Nalpha-Fmoc-Nomega,Nomega'-bis-Boc-D-arginine

  • MDL number MFCD00798643
  • Cas Number : 214852-34-3
  • Molecular Weight : 596.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-sup-i-n-i-sup-sup-bis-boc-d-arginine/3734?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-sup-i-n-i-sup-sup-bis-boc-d-arginine/3734?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart