CH6371379219 Chem Impex International, Inc.

Fmoc-D-citrulline

  • MDL number MFCD00151942
  • Cas Number : 200344-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-160 �C
Synonym Fmoc-D-Cit-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-citrulline/3740?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-citrulline/3740?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart