CH6371379220 Chem Impex International, Inc.

Fmoc-S-4-methylbenzyl-D-cysteine

  • MDL number MFCD00237013
  • Cas Number : 200354-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-157
Synonym Fmoc-D-Cys(pMeBzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-4-methylbenzyl-d-cysteine/3741?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-4-methylbenzyl-d-cysteine/3741?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart