CH6371379229 Chem Impex International, Inc.

Na-Fmoc-D-lysine hydrochloride

  • MDL number MFCD00190888
  • Cas Number : 201002-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-124 �C
Synonym Fmoc-D-Lys-OH HCl
Purity 99.8
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-d-lysine-hydrochloride/3751?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-d-lysine-hydrochloride/3751?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart