CH6371379232 Chem Impex International, Inc.

Fmoc-O-benzyl-D-threonine

  • MDL number MFCD00237033
  • Cas Number : 131545-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-113� C
Synonym Fmoc-D-Thr(Bzl)-OH
Purity 97
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-benzyl-d-threonine/3756?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-benzyl-d-threonine/3756?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart